Assay Details
Target Gene Details
Entrez Gene ID: | 71367 |
Gene Name: | carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9 |
Gene Aliases: |
5430438D01Rik |
Location: |
Chr.18:15451773-15760415 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chst9 | AK017407.1 |

Genomic Map