Assay Details
Target Gene Details
Entrez Gene ID: | 94184 |
Gene Name: | pyridoxal-dependent decarboxylase domain containing 1 |
Gene Aliases: |
2210010A19Rik, AA415817, Kiaa0251-hp |
Location: |
Chr.16:13833161-13903145 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdxdc1 | NM_001039533.2 | NP_001034622.1 | ||
| NM_001291017.1 | NP_001277946.1 | |||
| NM_053181.3 | 23 | 2931 | NP_444411.2 | |
| AK004611.1 | BAB23408.1 | |||
| AK030613.1 | 23 | 2762 | BAC27047.1 | |
| AK033107.1 | 23 | 2762 | BAC28155.1 | |
| AK054329.1 | BAC35732.1 | |||
| AK163696.1 | BAE37462.1 | |||
| AK165587.1 | 23 | 2766 | BAE38276.1 | |
| BC005541.1 | 23 | 2749 | AAH05541.1 | |
| BY469248.1 | 1 | 157 |

Genomic Map