Assay Details
Target Gene Details
Entrez Gene ID: | 224020 |
Gene Name: | phosphatidylinositol 4-kinase, catalytic, alpha polypeptide |
Gene Aliases: |
Pik4ca |
Location: |
Chr.16:17280351-17406314 on Build GRCm38 |
Assay Gene Location: | Within Exon 55 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pi4ka | NM_001001983.2 | 55 | 6338 | NP_001001983.2 |
| AK136942.1 | 19 | 2125 | BAE23184.1 | |
| BC049252.1 | 15 | 1676 | AAH49252.1 | |
| BC055479.1 | 9 | 1050 | AAH55479.1 | |
| BC075629.1 | 55 | 6338 | AAH75629.1 | |
| BU700531.1 | 1 | 112 |
More Information
Set Membership: |
|

Genomic Map