Assay Details
Target Gene Details
Entrez Gene ID: | 18596 |
Gene Name: | platelet derived growth factor receptor, beta polypeptide |
Gene Aliases: |
AI528809, CD140b, PDGFR-1, Pdgfr |
Location: |
Chr.18:61045150-61085068 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdgfrb | NM_001146268.1 | 3 | 574 | NP_001139740.1 |
| NM_008809.2 | 3 | 574 | NP_032835.2 | |
| AK134785.1 | 4 | 596 | BAE22283.1 | |
| AK143247.1 | 3 | 554 | ||
| BC055311.1 | 3 | 586 | AAH55311.1 | |
| BY753644.1 | 3 | 577 | ||
| X04367.1 | 3 | 283 | CAA27882.1 |
More Information
Set Membership: |
|

Genomic Map