Assay Details
Target Gene Details
Entrez Gene ID: | 68262 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acid acyltransferase, delta) |
Gene Aliases: |
1500003P24Rik |
Location: |
Chr.17:12118709-12219649 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agpat4 | NM_026644.2 | 9 | 1365 | NP_080920.2 |
| AK005139.1 | 9 | 1366 | BAB23837.2 | |
| AK020184.1 | ||||
| AK166917.1 | 9 | 1319 | BAE39115.1 | |
| BC047281.1 | 9 | 1296 | AAH47281.1 |
More Information
Set Membership: |
|

Genomic Map