Assay Details
Target Gene Details
Entrez Gene ID: | 107477 |
Gene Name: | guanylate cyclase activator 1B |
Gene Aliases: |
BC018258, GCAP2 |
Location: |
Chr.17:47384694-47392967 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Guca1b | NM_146079.1 | 4 | 934 | NP_666191.1 |
AK033052.1 | 4 | 899 | BAC28139.1 | |
BC018258.1 | 4 | 862 | AAH18258.1 | |
BC018480.1 | 4 | 903 | AAH18480.1 | |
BC029017.1 | 4 | 879 | AAH29017.1 | |
BC029661.1 | 4 | 889 | AAH29661.1 | |
BC031367.1 | 4 | 897 | AAH31367.1 | |
BC031916.1 | 4 | 934 | AAH31916.1 | |
BC033358.1 | 4 | 933 | AAH33358.1 | |
BC033380.1 | 4 | 889 | AAH33380.1 | |
BC033383.1 | 4 | 890 | AAH33383.1 | |
BC034331.1 | 4 | 890 | AAH34331.1 | |
BC034595.1 | 4 | 889 | AAH34595.1 |
