Assay Details
Target Gene Details
Entrez Gene ID: | 66648 |
Gene Name: | tubulin polyglutamylase complex subunit 2 |
Gene Aliases: |
5730437P09Rik, 5730494M16Rik, AI666318, Pgs2 |
Location: |
Chr.18:25127223-25169007 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpgs2 | NM_001004361.2 | 7 | 2994 | NP_001004361.2 |
| NM_001142697.1 | 7 | 2740 | NP_001136169.1 | |
| NM_001142698.1 | NP_001136170.1 | |||
| AK017627.1 | 1 | 871 | ||
| AK141632.1 | ||||
| AK171952.1 | 1 | 2160 | BAE42745.1 |
More Information
Set Membership: |
|

Genomic Map