Assay Details
Target Gene Details
Entrez Gene ID: | 27528 |
Gene Name: | neuronal regeneration related protein |
Gene Aliases: |
AI325076, D0H4S114, Harp, P311, PTZ17, SEZ17 |
Location: |
Chr.18:33437019-33464029 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrep | NM_001109988.1 | 5 | 1888 | NP_001103458.1 |
NM_001109989.2 | 5 | 1811 | NP_001103459.1 | |
NM_001267717.1 | 5 | 1800 | NP_001254646.1 | |
NM_053078.4 | 4 | 1926 | NP_444308.1 | |
AK048361.1 | 4 | 1726 | ||
AK133769.1 | 3 | 3021 | BAE21830.1 | |
BC054762.1 | 4 | 1708 | AAH54762.1 | |
D45203.1 | 1 | 1323 | ||
X70398.1 | 4 | 1722 | CAA49848.1 |
