Assay Details
Target Gene Details
Entrez Gene ID: | 11977 |
Gene Name: | ATPase, Cu++ transporting, alpha polypeptide |
Gene Aliases: |
MNK |
Location: |
Chr.X:106027224-106128160 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp7a | NM_001109757.2 | 23 | 5008 | NP_001103227.1 |
| NM_009726.5 | 23 | 5005 | NP_033856.3 | |
| AK030847.1 | 1 | 258 | ||
| AK040753.1 | 1 | 204 |
More Information
Set Membership: |
|

Genomic Map