Assay Details
Target Gene Details
Entrez Gene ID: | 245533 |
Gene Name: | acyl-CoA wax alcohol acyltransferase 1 |
Gene Aliases: |
Dgat2l3 |
Location: |
Chr.X:100572247-100578206 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Awat1 | NM_001081136.1 | 7 | 1155 | NP_001074605.1 |
| AK137763.1 | 7 | 1153 | ||
| BC147680.1 | 7 | 1126 | ||
| BC147683.1 | 7 | 1126 | ||
| BC147829.1 | 7 | 1084 | ||
| BC147832.1 | 7 | 1084 |
More Information
Set Membership: |
|

Genomic Map