Assay Details
Target Gene Details
Entrez Gene ID: | 68693 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U-like 2 |
Gene Aliases: |
1110031M08Rik, AI465155, Hnrpul2 |
Location: |
Chr.19:8819401-8834142 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hnrnpul2 | NM_001081196.1 | 1 | 239 | NP_001074665.1 |
Target Gene Details
Entrez Gene ID: | 70387 |
Gene Name: | tetratricopeptide repeat domain 9C |
Gene Aliases: |
2210019E14Rik, 6330408J23Rik |
Location: |
Chr.19:8809066-8819780 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc9c |
