Assay Details
Target Gene Details
| Entrez Gene ID: | 225888 | 
| Gene Name: | suppressor of variegation 4-20 homolog 1 (Drosophila) | 
| Gene Aliases: | AA117471, C630029K18Rik, KMT5B, Suv4-20h1 | 
| Location: | Chr.19:3764155-3818303 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Suv420h1 | NM_001167884.1 | 2 | 412 | NP_001161356.1 | 
| NM_001167885.1 | 2 | 412 | NP_001161357.1 | |
| NM_001167886.1 | 2 | 412 | NP_001161358.1 | |
| NM_001167887.1 | 2 | 412 | NP_001161359.1 | |
| NM_001167888.1 | 2 | 412 | NP_001161360.1 | |
| NM_001167889.1 | 2 | 412 | NP_001161361.1 | |
| NM_144871.4 | 2 | 412 | NP_659120.3 | |
| AK054093.1 | BAC35652.1 | |||
| AK082660.1 | 1 | 92 | BAC38565.1 | |
| AK083227.1 | 2 | 413 | BAC38817.1 | |
| AK087267.1 | 2 | 310 | BAC39834.1 | |
| AK139255.1 | BAE23934.1 | |||
| AY555192.1 | 1 | 102 | AAT00539.1 | |
| BC011214.1 | AAH11214.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic