Assay Details
Target Gene Details
Entrez Gene ID: | 12361 |
Gene Name: | calcium/calmodulin-dependent serine protein kinase (MAGUK family) |
Gene Aliases: |
DXPri1, DXRib1, LIN-2, Pals3, mLin-2 |
Location: |
Chr.X:13517080-13846784 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cask | NM_001284503.1 | NP_001271432.1 | ||
| NM_001284504.1 | NP_001271433.1 | |||
| NM_001284505.1 | NP_001271434.1 | |||
| NM_009806.3 | NP_033936.2 | |||
| BC141292.1 | ||||
| BC145620.1 | ||||
| Y17137.1 | CAA76646.1 | |||
| Y17138.1 | CAA76647.1 |
Target Gene Details
Entrez Gene ID: | 319200 |
Gene Name: | G protein-coupled receptor 82 |
Gene Aliases: |
- |
Location: |
Chr.X:13658153-13667433 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr82 | NM_175669.3 | 4 | 991 | NP_783600.1 |
| AK033761.1 | 4 | 993 | BAC28466.1 | |
| AY255571.1 | AAO85083.1 | |||
| BC116704.1 | 4 | 986 |
More Information
Set Membership: |
|

Genomic Map