Assay Details
Target Gene Details
Entrez Gene ID: | 245469 |
Gene Name: | PDZ domain containing 4 |
Gene Aliases: |
B230341P03, D130075J17Rik, Kiaa1444-hp, LU1, PDZRN4L, Pdzk4, Pdzrnk4l, Xlu |
Location: |
Chr.X:73793357-73824969 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdzd4 | NM_001029868.2 | 2 | 817 | NP_001025039.1 |
NM_001290540.1 | NP_001277469.1 | |||
AK040378.1 | 1 | 1247 | ||
AK046101.1 | ||||
AK084004.1 | 1 | 1319 | ||
AK165004.1 | 1 | 1401 | ||
BC056462.1 | 2 | 163 | AAH56462.1 |
