Assay Details
Target Gene Details
Entrez Gene ID: | 75404 |
Gene Name: | Rho GTPase activating protein 36 |
Gene Aliases: |
1100001E04Rik, rho-gap |
Location: |
Chr.X:49448328-49500250 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap36 | NM_001081123.1 | 13 | 2200 | NP_001074592.1 |
| AB073967.1 | 13 | 2067 | ||
| AK048167.1 | 13 | 2089 | ||
| BC090672.1 | 13 | 2171 |
More Information
Set Membership: |
|

Genomic Map