Assay Details
Target Gene Details
Entrez Gene ID: | 69860 |
Gene Name: | eukaryotic translation initiation factor 1A domain containing |
Gene Aliases: |
2010003J03Rik, AI131644 |
Location: |
Chr.19:5366813-5371511 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif1ad | NM_027236.2 | 6 | 1414 | NP_081512.1 |
| AK031807.1 | 6 | 1248 | ||
| AK158977.1 | 6 | 1232 | BAE34752.1 | |
| BC024640.1 | 6 | 1414 | AAH24640.1 |
More Information
Set Membership: |
|

Genomic Map