Assay Details
Target Gene Details
Entrez Gene ID: | 19265 |
Gene Name: | protein tyrosine phosphatase, receptor type, C polypeptide-associated protein |
Gene Aliases: |
AI605264, CD-45-AP, LPAP, LSM-1 |
Location: |
Chr.19:4154646-4156710 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprcap | NM_016933.3 | 2 | 587 | NP_058629.2 |
AI323196.1 | 2 | 559 | ||
AI627045.1 | 1 | 222 | ||
AK088334.1 | 2 | 585 | BAC40289.1 | |
AK165203.1 | 2 | 585 | BAE38076.1 | |
AW049152.1 | 1 | 276 | ||
BC013273.1 | 2 | 586 | AAH13273.1 | |
BC100560.1 | 2 | 568 | AAI00561.1 | |
D10105.1 | 2 | 563 | BAA00986.1 | |
U03856.1 | 1 | 580 | AAA67166.1 |
