Assay Details
Target Gene Details
Entrez Gene ID: | 210710 |
Gene Name: | growth factor receptor bound protein 2-associated protein 3 |
Gene Aliases: |
5930433H21Rik |
Location: |
Chr.X:74988545-75084905 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gab3 | NM_181584.4 | 4 | 918 | NP_853615.1 |
| AK031227.1 | 4 | 916 | BAC27308.1 | |
| AY057988.1 | 4 | 804 | AAL25824.1 | |
| BB634123.1 | ||||
| BC141240.1 | 4 | 798 |
More Information
Set Membership: |
|

Genomic Map