Assay Details
Target Gene Details
Entrez Gene ID: | 236852 |
Gene Name: | melanoma antigen family A, 10 |
Gene Aliases: |
- |
Location: |
Chr.X:72381870-72386858 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Magea10 | NM_001085506.1 | 4 | 778 | NP_001078975.1 |
| AK136064.1 | 4 | 779 |
More Information
Set Membership: |
|

Genomic Map