Assay Details
Target Gene Details
Entrez Gene ID: | 76705 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 2, pseudogene 7 |
Gene Aliases: |
1700023I07Rik |
Location: |
Chr.X:22461596-22463543 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r2-ps7 | NR_033731.1 | 1 | 410 | |
AK006280.1 | 1 | 377 | ||
AK133428.1 | 1 | 375 | BAE21653.1 |
