Assay Details
Target Gene Details
Entrez Gene ID: | 20589 |
Gene Name: | immunoglobulin mu binding protein 2 |
Gene Aliases: |
AEP, Catf1, RIPE3b1, Smbp-2, Smbp2, Smubp2, nmd, sma |
Location: |
Chr.19:3259076-3283010 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ighmbp2 | NM_009212.2 | 1 | 56 | NP_033238.2 |
| AK153763.1 | BAE32174.1 | |||
| AK154805.1 | 1 | 53 | BAE32841.1 | |
| AK155899.1 | 1 | 53 | BAE33491.1 | |
| BC066215.1 | AAH66215.1 | |||
| L10075.1 | AAA40143.1 |
Target Gene Details
Entrez Gene ID: | 353242 |
Gene Name: | mitochondrial ribosomal protein L21 |
Gene Aliases: |
BC028768 |
Location: |
Chr.19:3282920-3292837 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl21 |
More Information
Set Membership: |
|

Genomic Map