Assay Details
Target Gene Details
Entrez Gene ID: | 102902673 |
Gene Name: | predicted gene 21992 |
Gene Aliases: |
Rbm14-rbm4 |
Location: |
Chr.19:4784293-4811634 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21992 | NM_001290127.1 | 3 | 1005 | NP_001277056.1 |
| NM_001290128.1 | 2 | 582 | NP_001277057.1 | |
| EU287940.1 | 2 | 456 | ||
| EU287941.1 | 3 | 879 |
Target Gene Details
Entrez Gene ID: | 19653 |
Gene Name: | RNA binding motif protein 4 |
Gene Aliases: |
4921506I22Rik, Lark1, Mlark, Rbm4a, lark |
Location: |
Chr.19:4784293-4794009 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm4 | NM_001290122.1 | 3 | 736 | NP_001277051.1 |
| NM_001290123.1 | 3 | 725 | NP_001277052.1 | |
| NM_001290124.1 | NP_001277053.1 | |||
| NM_001290125.1 | NP_001277054.1 | |||
| NM_009032.3 | 3 | 772 | NP_033058.2 | |
| AK049659.1 | 3 | 733 | BAC33862.1 | |
| AK194488.1 | 1 | 106 | ||
| AK209057.1 | 1 | 106 | ||
| BC090633.1 | AAH90633.1 | |||
| BC130256.1 | 3 | 612 | ||
| BC144949.1 | 3 | 576 | ||
| BC144950.1 | ||||
| CK331622.1 | ||||
| DV660645.1 | ||||
| U89506.1 | 4 | 600 | AAC53171.1 |
More Information
Set Membership: |
|

Genomic Map