Assay Details
Target Gene Details
Entrez Gene ID: | 11605 |
Gene Name: | galactosidase, alpha |
Gene Aliases: |
Ags |
Location: |
Chr.X:134588169-134601005 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gla | NM_013463.2 | 7 | 2050 | NP_038491.2 |
| AK040081.1 | 7 | 1982 | BAC30508.1 | |
| AK164231.1 | 7 | 1983 | BAE37692.1 | |
| U34071.1 | 7 | 2000 | AAA96749.1 |
More Information
Set Membership: |
|

Genomic Map