Assay Details
Target Gene Details
Entrez Gene ID: | 226090 |
Gene Name: | endoplasmic reticulum metallopeptidase 1 |
Gene Aliases: |
D19Ertd410e, D19Wsu12e, Fxna, mKIAA1815 |
Location: |
Chr.19:29609883-29648484 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ermp1 | NM_001081213.1 | 15 | 4467 | NP_001074682.1 |
| AK129447.1 | 15 | 4395 | BAC98257.1 | |
| AK141229.1 | 1 | 960 | ||
| AK161868.1 | 1 | 1358 | ||
| BC031519.1 | 7 | 2743 | AAH31519.1 |
More Information
Set Membership: |
|

Genomic Map