Assay Details
Target Gene Details
Entrez Gene ID: | 26379 |
Gene Name: | estrogen related receptor, alpha |
Gene Aliases: |
ERRalpha, Err1, Estrra, Nr3b1 |
Location: |
Chr.19:6910977-6922048 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esrra | NM_007953.2 | 6 | 1025 | NP_031979.2 |
| AK019222.1 | 6 | 986 | ||
| AK156371.1 | 6 | 1001 | BAE33690.1 | |
| BC138586.1 | 6 | 985 | ||
| BC171958.1 | 6 | 985 | ||
| CO425338.1 | 5 | 675 | ||
| U85259.1 | 6 | 992 | AAB51250.1 |

Genomic Map