Assay Details
Target Gene Details
Entrez Gene ID: | 107250 |
Gene Name: | Kazal-type serine peptidase inhibitor domain 1 |
Gene Aliases: |
AI842353, Bono1, IGFBP-rP10 |
Location: |
Chr.19:45074485-45079289 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kazald1 | NM_178929.4 | 5 | 1312 | NP_849260.2 |
AK030678.1 | 5 | 1311 | BAC27075.1 | |
AK155095.1 | 6 | 1557 | BAE33042.1 | |
BX636599.1 | 1 | 72 |
