Assay Details
Target Gene Details
Entrez Gene ID: | 66704 |
Gene Name: | RNA binding motif protein 4B |
Gene Aliases: |
4921506I22Rik, AI504630, AI506404, Lark2, Neosin |
Location: |
Chr.19:4755146-4765940 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm4b | NM_025717.3 | 3 | 786 | NP_079993.2 |
| AK006447.1 | 3 | 769 | ||
| AK014823.1 | 3 | 710 | ||
| AK142116.1 | 3 | 741 | ||
| AK160796.1 | 3 | 1181 | ||
| AK181241.1 | 1 | 193 | ||
| AK182975.1 | 1 | 193 | ||
| BC019488.1 | 3 | 691 | AAH19488.1 |
More Information
Set Membership: |
|

Genomic Map