Assay Details
Target Gene Details
Entrez Gene ID: | 15185 |
Gene Name: | histone deacetylase 6 |
Gene Aliases: |
Hd6, Hdac5, Sfc6, mHDA2 |
Location: |
Chr.X:7930120-7947889 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac6 | NM_001130416.1 | 25 | 2948 | NP_001123888.1 |
NM_010413.3 | 25 | 3023 | NP_034543.3 | |
AF006603.2 | 24 | 2708 | AAD09835.2 | |
AK148092.1 | 25 | 2869 | ||
AK148144.1 | 25 | 2839 | BAE28372.1 | |
AK154099.1 | 25 | 2887 | BAE32376.1 | |
AK155545.1 | 23 | 3921 | ||
AK155546.1 | 25 | 2887 | ||
AK182664.1 | 1 | 75 | ||
AK184549.1 | 1 | 54 | ||
BC041105.1 | 25 | 2880 | AAH41105.1 |
