Assay Details
Target Gene Details
Entrez Gene ID: | 67395 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 2, pseudogene 9 |
Gene Aliases: |
4930403L05Rik |
Location: |
Chr.X:15110586-15111489 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r2-ps9 | NR_033171.1 | 1 | 435 | |
AK015065.1 | 1 | 440 | BAB29699.1 | |
AK015311.1 | 1 | 412 | BAB29790.1 | |
BC100448.1 | 1 | 412 | AAI00449.1 | |
BC132484.1 | 1 | 384 | ||
BC132486.1 | 1 | 384 |
