Assay Details
Target Gene Details
Entrez Gene ID: | 14156 |
Gene Name: | flap structure specific endonuclease 1 |
Gene Aliases: |
AW538437, FEN-1 |
Location: |
Chr.19:10199132-10204187 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fen1 | NM_001271614.1 | 2 | 1554 | NP_001258543.1 |
| NM_001271615.1 | 2 | 1449 | NP_001258544.1 | |
| NM_007999.4 | 2 | 1565 | NP_032025.2 | |
| AK042942.1 | 2 | 1398 | BAC31412.1 | |
| AK046256.1 | 2 | 1561 | BAC32660.1 | |
| AK076977.1 | 2 | 1539 | BAC36544.1 | |
| AK078778.1 | 2 | 1552 | BAC37390.1 | |
| AK153889.1 | 2 | 1532 | BAE32236.1 | |
| AK161047.1 | 2 | 1545 | BAE36169.1 | |
| AK166948.1 | 2 | 1533 | BAE39135.1 | |
| AK168731.1 | 2 | 1557 | BAE40572.1 | |
| BC010203.1 | 2 | 1495 | AAH10203.1 | |
| BC027295.1 | 2 | 1429 | AAH27295.1 | |
| L26320.1 | 1 | 1393 | AAC37664.1 |
More Information
Set Membership: |
|

Genomic Map