Assay Details
Target Gene Details
Entrez Gene ID: | 64697 |
Gene Name: | kidney expressed gene 1 |
Gene Aliases: |
0610008P16Rik, AI316519, GS4059 |
Location: |
Chr.19:12695790-12719902 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Keg1 | NM_029550.4 | 6 | 1109 | NP_083826.1 |
| AB028071.1 | 6 | 1086 | ||
| AK002360.1 | 6 | 1107 | BAB22039.1 | |
| AK165515.1 | 6 | 1107 | BAE38231.1 | |
| BC010803.1 | 6 | 1099 | AAH10803.1 |
More Information
Set Membership: |
|

Genomic Map