Assay Details
Target Gene Details
Entrez Gene ID: | 56613 |
Gene Name: | ribosomal protein S6 kinase, polypeptide 4 |
Gene Aliases: |
1110069D02Rik, 90kDa, AI848992, Msk2, mMSK2 |
Location: |
Chr.19:6829080-6841045 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps6ka4 | NM_019924.2 | 18 | 2691 | NP_064308.2 |
| AF074714.1 | 18 | 2663 | AAC67394.1 | |
| AK004412.1 | 8 | 1295 | ||
| AK089467.1 | 18 | 2685 | ||
| AK154674.1 | 18 | 2690 | BAE32757.1 | |
| BC012964.1 | 18 | 2701 | AAH12964.1 | |
| BF465098.1 | 2 | 400 | ||
| BP759267.1 | 1 | 487 |
More Information
Set Membership: |
|

Genomic Map