Assay Details
Target Gene Details
Entrez Gene ID: | 240613 |
Gene Name: | RIKEN cDNA 9930021J03 gene |
Gene Aliases: |
ENSMUSG00000048936, Gm9832, mKIAA2026 |
Location: |
Chr.19:29714402-29806106 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9930021J03Rik | NM_172836.3 | 8 | 6374 | NP_766424.2 |
| AK036879.1 | 1 | 1484 | BAC29616.1 | |
| AK047515.1 | 1 | 46 | ||
| AK220440.1 | 1 | 1705 | BAD90478.1 | |
| BC125582.1 | 1 | 1505 |

Genomic Map