Assay Details
Target Gene Details
Entrez Gene ID: | 269016 |
Gene Name: | SH3 domain containing ring finger 2 |
Gene Aliases: |
2310046K19, 9130023G24Rik, AI452083, Ppp1r39, RNF158 |
Location: |
Chr.18:42051743-42158695 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3rf2 | NM_001146299.1 | 2 | 668 | NP_001139771.1 |
| NM_172966.3 | 2 | 668 | NP_766554.2 | |
| AK028153.1 | 1 | 68 | BAC25780.1 | |
| AK029168.1 | 2 | 669 | BAC26334.1 | |
| AK033601.1 | 2 | 712 | BAC28382.1 | |
| BC137955.1 | 2 | 621 | ||
| BC145060.1 | 2 | 621 |

Genomic Map