Assay Details
Target Gene Details
Entrez Gene ID: | 66745 |
Gene Name: | tumor protein D52-like 3 |
Gene Aliases: |
4931412G03Rik |
Location: |
Chr.19:30003790-30006020 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpd52l3 | NM_025741.2 | 1 | 742 | NP_080017.1 |
| AK016362.1 | 1 | 731 | BAB30205.1 | |
| AK016450.1 | 1 | 742 | BAB30244.1 | |
| BC125314.1 | 1 | 742 | ||
| BC125316.1 | 1 | 742 |

Genomic Map