Assay Details
Target Gene Details
Entrez Gene ID: | 68041 |
Gene Name: | Mid1 interacting protein 1 (gastrulation specific G12-like (zebrafish)) |
Gene Aliases: |
3110038L01Rik, Mig12, S14R |
Location: |
Chr.X:10717365-10719702 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mid1ip1 | NM_001166635.1 | 2 | 619 | NP_001160107.1 |
NM_026524.4 | 2 | 616 | NP_080800.1 | |
AK004021.1 | 1 | 120 | BAB23129.1 | |
AK014143.1 | 2 | 395 | BAB29178.1 | |
AK090003.1 | 2 | 502 | BAC41039.1 | |
AK131619.1 | 2 | 395 | BAE20724.1 | |
AK151153.1 | 2 | 478 | BAE30159.1 | |
AK159166.1 | 2 | 500 | BAE34868.1 | |
AK160062.1 | 2 | 544 | BAE35598.1 | |
AK172143.1 | 2 | 503 | BAE42845.1 | |
AY263385.1 | 1 | 247 | AAP87014.1 | |
BC004014.1 | 2 | 499 | AAH04014.1 | |
BC010778.1 | 1 | 34 | AAH10778.1 | |
BC052899.1 | 2 | 494 | AAH52899.1 | |
CT010221.1 | CAJ18429.1 |
