Assay Details
Target Gene Details
Entrez Gene ID: | 226025 |
Gene Name: | transient receptor potential cation channel, subfamily M, member 3 |
Gene Aliases: |
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2 |
Location: |
Chr.19:22138181-22989897 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trpm3 | NM_001035239.2 | NP_001030316.1 | ||
NM_001035240.2 | NP_001030317.1 | |||
NM_001035241.2 | NP_001030318.1 | |||
NM_001035242.1 | NP_001030319.1 | |||
NM_001035243.2 | NP_001030320.1 | |||
NM_177341.4 | 1 | 213 | NP_796315.2 | |
AJ544532.1 | CAD67492.1 | |||
AJ544533.1 | CAD67493.1 | |||
AJ544534.1 | CAD67494.1 | |||
AJ544535.1 | CAD67495.1 | |||
AJ544536.1 | CAD67496.1 | |||
AK044454.1 | 1 | 215 | BAC31929.1 | |
AK173218.1 | BAD32496.1 | |||
BC141400.1 | 1 | 189 | ||
BC141412.1 | 1 | 189 | ||
HM064407.1 | 1 | 238 | ||
HM064408.1 | 1 | 238 | ||
HM064409.1 | 1 | 238 | ||
HM064410.1 | 1 | 238 | ||
HM064411.1 | 1 | 238 | ||
JF706722.1 | 1 | 209 | ||
JX644970.1 | 1 | 238 | ||
JX644971.1 | 1 | 238 | ||
JX644972.1 | 1 | 238 | ||
JX644973.1 | 1 | 238 | ||
JX644974.1 | 1 | 238 | ||
JX644975.1 | 1 | 238 | ||
JX644976.1 | 1 | 238 | ||
JX644977.1 | 1 | 238 | ||
JX644978.1 | 1 | 238 | ||
JX644979.1 | 1 | 238 | ||
JX644980.1 | 1 | 238 | ||
JX644981.1 | 1 | 238 | ||
JX644982.1 | 1 | 238 | ||
JX644983.1 | 1 | 238 | ||
JX644984.1 | 1 | 238 | ||
JX644985.1 | 1 | 238 | ||
JX644986.1 | 1 | 238 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
