Assay Details
Target Gene Details
Entrez Gene ID: | 209268 |
Gene Name: | immunoglobulin superfamily, member 1 |
Gene Aliases: |
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364 |
Location: |
Chr.X:49782536-49797906 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf1 | NM_177591.4 | NP_808259.2 | ||
| NM_177915.4 | 5 | 1355 | NP_808583.1 | |
| NM_183335.2 | NP_899178.2 | |||
| AK030452.1 | 5 | 1327 | BAC26969.1 | |
| AK129126.1 | BAC97936.1 | |||
| AY227771.1 | AAP57079.1 | |||
| AY227772.1 | 5 | 1349 | AAP57080.1 | |
| AY227773.1 | AAP57081.1 | |||
| BC089474.1 | 5 | 1336 | AAH89474.1 | |
| BC100420.1 | 5 | 1355 | AAI00421.1 |
More Information
Set Membership: |
|

Genomic Map