Assay Details
Target Gene Details
Entrez Gene ID: | 209268 |
Gene Name: | immunoglobulin superfamily, member 1 |
Gene Aliases: |
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364 |
Location: |
Chr.X:49782536-49797906 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf1 | NM_177591.4 | 13 | 2271 | NP_808259.2 |
| NM_183335.2 | 12 | 2135 | NP_899178.2 | |
| NM_183336.2 | 4 | 525 | NP_899179.1 | |
| AK030701.1 | 4 | 527 | ||
| AK076427.1 | 4 | 518 | BAC36338.1 | |
| AK129126.1 | 13 | 2253 | BAC97936.1 | |
| AY227771.1 | 13 | 2265 | AAP57079.1 | |
| AY227773.1 | 12 | 2135 | AAP57081.1 | |
| AY227774.1 | 4 | 520 | AAP57082.1 | |
| BC119264.1 | 4 | 488 | ||
| BC119266.1 | 4 | 488 |
More Information
Set Membership: |
|

Genomic Map