Assay Details
Target Gene Details
Entrez Gene ID: | 26403 |
Gene Name: | mitogen-activated protein kinase kinase kinase 11 |
Gene Aliases: |
2610017K16Rik, Mlk3, PTK1, SPRK |
Location: |
Chr.19:5688740-5702865 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k11 |
Target Gene Details
Entrez Gene ID: | 104401 |
Gene Name: | pecanex-like 3 (Drosophila) |
Gene Aliases: |
mKIAA4073 |
Location: |
Chr.19:5664635-5688908 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcnxl3 | NM_144868.3 | 1 | 148 | NP_659117.2 |

Genomic Map