Assay Details
Target Gene Details
Entrez Gene ID: | 107328 |
Gene Name: | tRNA phosphotransferase 1 |
Gene Aliases: |
AU079016, AW045591, Tpt1, Tpt1h |
Location: |
Chr.19:6996131-6999048 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpt1 | NM_153597.2 | 1 | 319 | NP_705825.2 |
| NR_027351.1 | 1 | 319 | ||
| AK038830.1 | 1 | 321 | ||
| AK090183.1 | 1 | 292 | ||
| BC027575.1 | 1 | 117 | AAH27575.1 |
More Information
Set Membership: |
|

Genomic Map