Assay Details
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcx | NM_001162946.1 | 21 | 3667 | NP_001156418.1 |
| NM_008797.3 | 20 | 3644 | NP_032823.2 | |
| AK077665.1 | 12 | 2472 | BAC36939.1 | |
| AK148328.1 | 21 | 3614 | BAE28484.1 | |
| AK170596.1 | 21 | 3627 | BAE41902.1 | |
| AK170871.1 | 21 | 3618 | ||
| AK172320.1 | 21 | 3595 | BAE42943.1 | |
| BC055030.1 | 21 | 3602 | AAH55030.1 | |
| L09192.1 | 20 | 3620 | AAA39737.1 | |
| M97957.1 | 15 | 2792 | AAA39897.1 |
More Information
Set Membership: |
|

Genomic Map