Assay Details
Target Gene Details
Entrez Gene ID: | 69534 |
Gene Name: | arginine vasopressin-induced 1 |
Gene Aliases: |
2310008N12Rik, AA410041, VIT32, esu, mVIT32 |
Location: |
Chr.19:42123275-42128993 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Avpi1 | NM_027106.4 | 3 | 875 | NP_081382.1 |
| AK009243.1 | 3 | 876 | BAB26163.1 | |
| AK177091.1 | ||||
| AY505175.1 | 1 | 249 | ||
| BC027646.1 | AAH27646.1 |

Genomic Map