Assay Details
Target Gene Details
Entrez Gene ID: | 108902 |
Gene Name: | beta-1,4-glucuronyltransferase 1 |
Gene Aliases: |
1500032M01Rik, B3gnt1, B3gnt6, BETA3GNT1, iGAT, iGNT |
Location: |
Chr.19:5038826-5041134 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4gat1 | NM_175383.2 | 1 | 202 | NP_780592.1 |
| AK050347.1 | 1 | 176 | BAC34203.1 | |
| AK075733.1 | 1 | 83 | ||
| AK083857.1 | 1 | 125 | BAC39041.1 | |
| AK158899.1 | 1 | 203 | BAE34720.1 | |
| BC069927.1 | 1 | 130 | AAH69927.1 |
More Information
Set Membership: |
|

Genomic Map