Assay Details
Target Gene Details
Entrez Gene ID: | 74931 |
Gene Name: | RIKEN cDNA 4930481A15 gene |
Gene Aliases: |
- |
Location: |
Chr.19:5406815-5422847 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930481A15Rik | NR_015545.2 | |||
NR_027938.2 | ||||
AK015604.1 | ||||
AK039982.1 | ||||
BI202508.1 | ||||
BM937611.1 |
