Assay Details
Target Gene Details
Entrez Gene ID: | 12015 |
Gene Name: | BCL2-associated agonist of cell death |
Gene Aliases: |
AI325008, Bbc2 |
Location: |
Chr.19:6941861-6951905 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bad | NM_007522.3 | 1 | 481 | NP_031548.1 |
BC006762.1 | 1 | 449 | AAH06762.1 | |
L37296.1 | 1 | 489 | AAA64465.1 |
Target Gene Details
Entrez Gene ID: | 107173 |
Gene Name: | G protein-coupled receptor 137 |
Gene Aliases: |
AI428855 |
Location: |
Chr.19:6938062-6942450 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpr137 | NM_001177361.1 | 1 | 110 | NP_001170832.1 |
AK143450.1 | 1 | 111 | BAE25382.1 |
