Assay Details
Target Gene Details
Entrez Gene ID: | 74481 |
Gene Name: | basic leucine zipper transcription factor, ATF-like 2 |
Gene Aliases: |
4933430F08Rik |
Location: |
Chr.19:6164392-6172476 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Batf2 | NM_028967.1 | 3 | 1173 | NP_083243.1 |
| AK150133.1 | BAE29330.1 | |||
| AK171828.1 | 3 | 1190 | BAE42686.1 | |
| BC024521.1 | 3 | 1173 | AAH24521.1 |
More Information
Set Membership: |
|

Genomic Map