Assay Details
Target Gene Details
Entrez Gene ID: | 170459 |
Gene Name: | StAR-related lipid transfer (START) domain containing 4 |
Gene Aliases: |
4632419C16Rik, 9030213J02Rik, AA517649, AW324468 |
Location: |
Chr.18:33199140-33213879 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stard4 | NM_133774.4 | 6 | 2585 | NP_598535.1 |
| AK014587.1 | 1 | 2381 | ||
| AK048754.1 | 1 | 624 | ||
| AK054501.1 | 6 | 2586 | BAC35805.1 | |
| AK085331.1 | 6 | 2589 | BAC39424.1 |

Genomic Map