Assay Details
Target Gene Details
Entrez Gene ID: | 22289 |
Gene Name: | lysine (K)-specific demethylase 6A |
Gene Aliases: |
Utx |
Location: |
Chr.X:18162533-18279791 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm6a | NM_009483.1 | 17 | 2386 | NP_033509.1 |
| AJ002730.1 | 16 | 1845 | CAA05692.1 | |
| AK080370.1 | 17 | 2405 | ||
| AK144654.1 | 15 | 2222 | ||
| BC027130.1 | 5 | 833 | AAH27130.1 | |
| BC053433.1 | 17 | 2386 | AAH53433.1 | |
| BC075703.1 | 8 | 1296 | AAH75703.1 |
More Information
Set Membership: |
|

Genomic Map