Assay Details
Target Gene Details
Entrez Gene ID: | 52808 |
Gene Name: | TSPY-like 2 |
Gene Aliases: |
CDA1, CINAP, DENTT, DXBwg1396e, DXHXS1008E, E130307F10Rik |
Location: |
Chr.X:152336852-152342506 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspyl2 | NM_029836.3 | 1 | 315 | NP_084112.1 |
| AK018700.1 | 1 | 257 | BAB31351.1 | |
| AY273809.1 | 1 | 156 | AAQ17208.1 | |
| BC054393.1 | 1 | 235 | AAH54393.1 | |
| CJ066847.1 |
More Information
Set Membership: |
|

Genomic Map